Assay Details
| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106023.1 | NP_001099493.2 | 10 - 11 | 1168 | 73 | |
| XM_006251615.2 | XP_006251677.1 | 10 - 11 | 1486 | 73 | ||
| GenBank mRNA | FQ232613.1 | - | 9 - 10 | 907 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erlec1
Genomic Map