Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135169.2 | NP_001128641.1 | 3 - 4 | 229 | 116 | |
| NM_134395.2 | NP_599222.2 | 2 - 3 | 442 | 116 | ||
| XM_006253777.2 | XP_006253839.1 | 3 - 4 | 432 | 116 | ||
| GenBank mRNA | AF083396.1 | - | 2 - 3 | 450 | 116 | |
| BC158854.1 | - | 2 - 3 | 123 | 7936967 | 116 | |
| CB744788.1 | - | 3 - 4 | 283 | 116 | ||
| CV107072.1 | - | 3 - 4 | 116 | |||
| FQ211234.1 | - | 3 - 4 | 219 | 116 | ||
| FQ217516.1 | - | 3 - 4 | 219 | 116 | ||
| FQ218209.1 | - | 3 - 4 | 225 | 116 | ||
| FQ223586.1 | - | 3 - 4 | 209 | 116 | ||
| FQ223742.1 | - | 3 - 4 | 219 | 116 | ||
| FQ224495.1 | - | 3 - 4 | 225 | 116 | ||
| FQ225707.1 | - | 3 - 4 | 199 | 116 | ||
| FQ230083.1 | - | 3 - 4 | 247 | 116 | ||
| FQ230323.1 | - | 3 - 4 | 228 | 116 | ||
| FQ231364.1 | - | 3 - 4 | 210 | 115 | ||
| FQ233821.1 | - | 3 - 4 | 217 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmem14c
Genomic Map