Assay Details
| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108407.1 | NP_001101877.1 | 6 - 7 | 516 | 64 | |
| XM_006253674.2 | XP_006253736.1 | 6 - 7 | 614 | 64 | ||
| XM_006253675.2 | XP_006253737.1 | 5 - 6 | 524 | 64 | ||
| XM_008771499.1 | XP_008769721.1 | 6 - 7 | 1570 | 64 | ||
| GenBank mRNA | FQ218844.1 | - | 6 - 7 | 608 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Auh
Genomic Map