Assay Details
| Gene Symbol: Phf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107342.1 | NP_001100812.1 | 4 - 5 | 582 | 61 | |
| XM_006253729.2 | XP_006253791.1 | 5 - 6 | 2604 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P464092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176259 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P464092 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P464092 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P464092 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P206170 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26447 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15449 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10794418 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794422 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794419 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794426 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794424 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794415 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794427 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794425 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794423 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794410 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794412 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794420 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794421 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794417 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794428 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794429 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10794409 | - | - |
| Rat Genome 230 2.0 Array | 1390103_at | - | - |
| Rat Genome 230 2.0 Array | 1390103_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390103_at | - | - |
| Affymetrix HT Rat Focus Array | 1390103_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700002294.rn.1 | TC1700000289.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390103_PM_at | - | - |
| Rat Expression Set 230 | 1390103_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794409 | - | - |
| Rat Genome U34 Set | rc_AI169319_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22412515 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355601 | - | - |

Phf2
Genomic Map