Assay Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005542.2 | NP_001005542.2 | 4 - 5 | 715 | 80 | |
| GenBank mRNA | BC082103.1 | - | 3 - 4 | 499 | 7134619 | 80 |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr34
Genomic Map