Assay Details
| Gene Symbol: Gtf3c4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109473.1 | NP_001102943.1 | 2 - 3 | 2287 | 96 | |
| XM_006233872.2 | XP_006233934.1 | 2 - 3 | 3064 | 96 | ||
| XM_008761680.1 | XP_008759902.1 | 3 - 4 | 2012 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c4
Genomic Map