Assay Details
| Gene Symbol: Gtf3c4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109473.1 | NP_001102943.1 | 3 - 4 | 2415 | 103 | |
| XM_006233872.2 | XP_006233934.1 | 3 - 4 | 3192 | 103 | ||
| XM_008761680.1 | XP_008759902.1 | 4 - 5 | 2140 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c4
Genomic Map