Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237919.2 | XP_006237981.1 | 2 - 3 | 517 | 125 | |
| XM_232798.8 | XP_232798.4 | 2 - 3 | 347 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006237919.2 transcripts.
Set Membership: |
|

Osgin2
Genomic Map