Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237919.2 | XP_006237981.1 | 5 - 6 | 932 | 78 | |
| XM_232798.8 | XP_232798.4 | 5 - 6 | 762 | 78 | ||
More Information
Set Membership: |
|

Osgin2
Genomic Map