Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033705.1 | NP_001028877.1 | 6 - 7 | 734 | 88 | |
| GenBank mRNA | BC101930.1 | - | 6 - 7 | 734 | 7938755 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp451
Genomic Map