Assay Details
Gene Symbol: Cnot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108794.2 | NP_001102264.1 | 5 - 6 | 1289 | 88 | |
GenBank mRNA | BC158750.1 | - | 5 - 6 | 1289 | 9101674 | 88 |
FQ233266.1 | - | 5 - 6 | 1294 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
