Assay Details
| Gene Symbol: RGD1308742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234051.2 | XP_006234113.2 | 5 - 6 | 665 | 107 | |
| XM_008761799.1 | XP_008760021.1 | 3 - 4 | 431 | 107 | ||
| XM_008761800.1 | XP_008760022.1 | 3 - 4 | 428 | 107 | ||
| XM_342422.5 | XP_342423.5 | 5 - 6 | 662 | 107 | ||
More Information
Set Membership: |
|

RGD1308742
Genomic Map