Assay Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024344.1 | NP_001019515.1 | 2 - 3 | 292 | 98 | |
| GenBank mRNA | BC088199.1 | - | 2 - 3 | 292 | 7377221 | 98 |
| FQ225852.1 | - | 1 - 2 | 207 | 98 | ||
| FQ233921.1 | - | 1 - 2 | 207 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sit1
Genomic Map