Assay Details
| Gene Symbol: Riok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009687.1 | NP_001009687.1 | 4 - 5 | 512 | 68 | |
| GenBank mRNA | BC088297.1 | - | 4 - 5 | 512 | 7322759 | 68 |
| FQ232861.1 | - | 4 - 5 | 547 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Riok2
Genomic Map