Assay Details
| Gene Symbol: Prelid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009636.1 | NP_001009636.1 | 3 - 4 | 570 | 69 | |
| GenBank mRNA | BC088284.1 | - | 3 - 4 | 570 | 7323653 | 69 |
More Information
Set Membership: |
|

Prelid1
Genomic Map