Assay Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198781.2 | NP_942076.2 | 1 - 2 | 51 | 92 | |
| XM_006235944.2 | XP_006236006.1 | 1 - 2 | 787 | 92 | ||
| GenBank mRNA | DY313123.1 | - | 1 - 2 | 51 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_198781.2 transcripts.
Set Membership: |
|

Tmub1
Genomic Map