Assay Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080153.1 | NP_001073622.1 | 2 - 3 | 558 | 74 | |
| NM_198781.2 | NP_942076.2 | 2 - 3 | 468 | 74 | ||
| XM_006235944.2 | XP_006236006.1 | 2 - 3 | 1204 | 74 | ||
| XM_006235945.2 | XP_006236007.1 | 2 - 3 | 595 | 74 | ||
| XM_008762651.1 | XP_008760873.1 | 2 - 3 | 470 | 74 | ||
| GenBank mRNA | AY603378.1 | - | 2 - 3 | 558 | 74 | |
| DY313123.1 | - | 2 - 3 | 468 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmub1
Genomic Map