Assay Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251199.1 | XP_006251261.1 | 1 - 2 | 1731 | 139 | |
| XM_006251200.2 | XP_006251262.1 | 2 - 3 | 1084 | 139 | ||
More Information
Set Membership: |
|

Osm
Genomic Map