Assay Details
| Gene Symbol: Senp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105888.1 | NP_001099358.1 | 19 - 20 | 2787 | 64 | |
| XM_006248244.2 | XP_006248306.1 | 19 - 20 | 3742 | 64 | ||
| XM_006248247.2 | XP_006248309.1 | 18 - 19 | 3545 | 64 | ||
| XM_006248248.2 | XP_006248310.1 | 17 - 18 | 3352 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Senp7
Genomic Map