Assay Details
| Gene Symbol: Neurl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107605.1 | NP_001101075.1 | 3 - 4 | 713 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translation
protein monoubiquitination
sperm axoneme assembly
brain development
skeletal muscle tissue development
lactation
negative regulation of cell proliferation
sperm motility
positive regulation of apoptotic process
positive regulation of epidermal growth factor-activated receptor activity
negative regulation of Notch signaling pathway
positive regulation of long-term neuronal synaptic plasticity
positive regulation of filopodium assembly
positive regulation of dendritic spine development
cellular response to amino acid stimulus
positive regulation of synapse maturation
molecular_function
ubiquitin-protein transferase activity
zinc ion binding
translation factor activity, non-nucleic acid binding
ubiquitin protein ligase activity
regulation of translation
protein monoubiquitination
sperm axoneme assembly
brain development
skeletal muscle tissue development
lactation
negative regulation of cell proliferation
sperm motility
positive regulation of apoptotic process
positive regulation of epidermal growth factor-activated receptor activity
negative regulation of Notch signaling pathway
positive regulation of long-term neuronal synaptic plasticity
positive regulation of filopodium assembly
positive regulation of dendritic spine development
cellular response to amino acid stimulus
positive regulation of synapse maturation
molecular_function
ubiquitin-protein transferase activity
zinc ion binding
translation factor activity, non-nucleic acid binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P084684 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P084683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5660 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084683 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084684 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084683 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084684 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P084684 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P084683 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P506221 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78376 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10715955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715956 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10715952 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100030780.rn.1 | TC0100003765.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10715952 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650627 | - | - |

Neurl1
Genomic Map