Assay Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012275.2 | NP_001012275.1 | 6 - 7 | 722 | 61 | |
| XM_006243784.2 | XP_006243846.1 | 6 - 7 | 1190 | 61 | ||
| XM_006243785.2 | XP_006243847.1 | 6 - 7 | 1187 | 61 | ||
| XM_006243787.2 | XP_006243849.1 | 5 - 6 | 411 | 61 | ||
| XM_008766537.1 | XP_008764759.1 | 6 - 7 | 875 | 61 | ||
| XM_008766538.1 | XP_008764760.1 | 6 - 7 | 878 | 61 | ||
| GenBank mRNA | CV126331.1 | - | 2 - 3 | 226 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arih2
Genomic Map