Assay Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244005.1 | XP_006244067.1 | 9 - 10 | 1425 | 54 | |
| XM_006244006.2 | XP_006244068.1 | 9 - 10 | 2081 | 54 | ||
| XM_006244007.1 | XP_006244069.1 | 9 - 10 | 1239 | 54 | ||
| XM_236647.7 | XP_236647.6 | 9 - 10 | 1410 | 54 | ||
More Information
Set Membership: |
|

Klhl18
Genomic Map