Assay Details
| Gene Symbol: Lars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108787.1 | NP_001102257.1 | 12 - 13 | 1752 | 57 | |
| XM_006244201.2 | XP_006244263.1 | 14 - 15 | 1979 | 57 | ||
| XM_008766727.1 | XP_008764949.1 | 12 - 13 | 1826 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lars2
Genomic Map