Assay Details
| Gene Symbol: Lca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013954.1 | NP_001013976.1 | 3 - 4 | 552 | 80 | |
| XM_008766249.1 | XP_008764471.1 | 2 - 3 | 496 | 80 | ||
| XM_008766250.1 | XP_008764472.1 | 1 - 2 | 377 | 80 | ||
| XM_008766251.1 | XP_008764473.1 | 3 - 4 | 587 | 80 | ||
| XM_008766252.1 | XP_008764474.1 | 2 - 3 | 440 | 80 | ||
| XM_008766253.1 | XP_008764475.1 | 2 - 3 | 883 | 80 | ||
| XM_008766255.1 | XP_008764477.1 | 3 - 4 | 612 | 80 | ||
| XM_008766256.1 | XP_008764478.1 | 2 - 3 | 418 | 80 | ||
| XM_008766257.1 | XP_008764479.1 | 2 - 3 | 1057 | 80 | ||
| XM_008766258.1 | XP_008764480.1 | 2 - 3 | 4081 | 80 | ||
| XM_008766259.1 | XP_008764481.1 | 2 - 3 | 497 | 80 | ||
| GenBank mRNA | BC083657.1 | - | 1 - 2 | 375 | 80 | |
| BC085805.1 | - | 3 - 4 | 552 | 7112021 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lca5
Genomic Map