Assay Details
| Gene Symbol: Dnajc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750579.2 | XP_003750627.1 | 17 - 18 | 2144 | 58 | |
| XM_008766592.1 | XP_008764814.1 | 16 - 17 | 1908 | 58 | ||
| XM_008766593.1 | XP_008764815.1 | 17 - 18 | 2144 | 58 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P445903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101455 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P314398 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P445903 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P147622 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2233 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10919742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919717 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919771 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919763 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919748 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919743 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919718 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919740 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919747 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919730 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919773 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919761 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919762 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919722 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919770 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919764 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919767 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919751 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919757 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919746 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919750 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919737 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919768 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919755 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919724 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919765 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919760 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919749 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919756 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919754 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919759 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919772 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919726 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919745 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919744 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919753 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919752 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919766 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919758 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919769 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10919716 | - | - |
| Rat Genome 230 2.0 Array | 1372189_at | - | - |
| Rat Genome 230 2.0 Array | 1372189_PM_at | - | - |
| Rat Expression Set 230 | 1372189_at | - | - |
| Rat Expression Set 230 | 1372189_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919716 | - | - |
| Rat Genome U34 Set | rc_AA943029_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21311961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358984 | - | - |

Dnajc13
Genomic Map