Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100725.1 | NP_001094195.1 | 4 - 5 | 426 | 60 | |
| GenBank mRNA | BC089995.1 | - | 3 - 4 | 317 | 60 | |
| FQ226090.1 | - | 4 - 5 | 431 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Exosc7
Genomic Map