Assay Details
| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023962.2 | NP_076452.1 | 2 - 3 | 443 | 75 | |
| GenBank mRNA | AB052170.2 | - | 2 - 3 | 443 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfd
Genomic Map