Assay Details
| Gene Symbol: Tex10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106653.1 | NP_001100123.1 | 10 - 11 | 2514 | 62 | |
| XM_006238034.2 | XP_006238096.1 | 9 - 10 | 3540 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex10
Genomic Map