Assay Details
| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175581.3 | NP_783171.2 | 4 - 5 | 470 | 72 | |
| XM_008771434.1 | XP_008769656.1 | 3 - 4 | 391 | 72 | ||
| GenBank mRNA | BC097484.1 | - | 4 - 5 | 470 | 7459242 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Ctsr
Genomic Map