Assay Details
| Gene Symbol: RGD1564053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109259.2 | NP_001102729.2 | 4 - 5 | 924 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1564053
Genomic Map