Assay Details
| Gene Symbol: Ubxn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107905.1 | NP_001101375.1 | 3 - 4 | 363 | 85 | |
| XM_006237837.2 | XP_006237899.1 | 3 - 4 | 381 | 85 | ||
| XM_008763540.1 | XP_008761762.1 | 2 - 3 | 256 | 85 | ||
| XM_008763541.1 | XP_008761763.1 | 3 - 4 | 381 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn2b
Genomic Map