Assay Details
| Gene Symbol: Agk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127497.1 | NP_001120969.1 | 15 - 16 | 1178 | 58 | |
| XM_006236363.2 | XP_006236425.1 | 15 - 16 | 1480 | 58 | ||
| XM_008762821.1 | XP_008761043.1 | 15 - 16 | 1575 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P153338 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P492808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154959 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153338 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P492808 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153338 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P492808 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P492808 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P153338 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P492808 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P265163 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10854697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854706 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854703 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10854689 | - | - |
| Rat Genome 230 2.0 Array | 1373925_at | - | - |
| Rat Genome 230 2.0 Array | 1373925_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373925_at | - | - |
| Affymetrix HT Rat Focus Array | 1373925_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400005159.rn.1 | TC0400000698.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400005158.rn.1 | TC0400000698.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373925_at | - | - |
| Rat Expression Set 230 | 1373925_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854689 | - | - |
| Rat Genome U34 Set | rc_AI113029_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22076883 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650198 | - | - |

Agk
Genomic Map