Assay Details
| Gene Symbol: Arid3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108066.2 | NP_001101536.2 | 6 - 7 | 1520 | 68 | |
| XM_006240902.2 | XP_006240964.1 | 6 - 7 | 1835 | 68 | ||
| XM_006240903.1 | XP_006240965.1 | 6 - 7 | 1653 | 68 | ||
| XM_006240904.2 | XP_006240966.1 | 6 - 7 | 1655 | 68 | ||
| XM_006240905.2 | XP_006240967.1 | 6 - 7 | 2018 | 68 | ||
| XM_008765169.1 | XP_008763391.1 | 6 - 7 | 1526 | 68 | ||
| GenBank mRNA | BC168846.1 | - | 6 - 7 | 1520 | 7114467 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Arid3a
Genomic Map