Assay Details
| Gene Symbol: March5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106372.2 | NP_001099842.1 | 2 - 3 | 526 | 114 | |
| XM_008760367.1 | XP_008758589.1 | 2 - 3 | 532 | 114 | ||
| GenBank mRNA | BC158801.1 | - | 2 - 3 | 526 | 7321043 | 114 |
More Information
Updated Mapping Information
Set Membership: |
|

March5
Genomic Map