Assay Details
| Gene Symbol: Pdcd1lg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107582.2 | NP_001101052.2 | 3 - 4 | 604 | 67 | |
| XM_006231209.2 | XP_006231271.1 | 2 - 3 | 656 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P128032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149501 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P128032 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P128032 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P128032 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P177980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10714701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714703 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10714700 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100028007.rn.1 | TC0100003447.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100028006.rn.1 | TC0100003447.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100028005.rn.1 | TC0100003447.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100028009.rn.1 | TC0100003447.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100028010.rn.1 | TC0100003447.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10714700 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21247420 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2039016 | - | - |

Pdcd1lg2
Genomic Map