Assay Details
| Gene Symbol: Efna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_574979.5 | XP_574979.4 | 2 - 3 | 419 | 64 | |
| GenBank mRNA | AY045577.1 | - | 1 - 2 | 233 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Efna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_574979.5 | XP_574979.4 | 2 - 3 | 419 | 64 | |
| GenBank mRNA | AY045577.1 | - | 1 - 2 | 233 | 64 | |
Set Membership: |
|