Assay Details
| Gene Symbol: Zfp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168642.1 | NP_001162113.1 | 5 - 6 | 589 | 133 | |
| XM_006228106.2 | XP_006228168.1 | 5 - 6 | 632 | 133 | ||
| XM_008758775.1 | XP_008756997.1 | 5 - 6 | 648 | 133 | ||
| XM_008758776.1 | XP_008756998.1 | 3 - 4 | 273 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp40
Genomic Map