Assay Details
| Gene Symbol: Car4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019174.3 | NP_062047.1 | 5 - 6 | 611 | 58 | |
| GenBank mRNA | BC097329.1 | - | 5 - 6 | 506 | 7445156 | 58 |
| CB746957.1 | - | 2 - 3 | 172 | 58 | ||
| FQ222059.1 | - | 561 | 58 | |||
| S68245.1 | - | 5 - 6 | 554 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car4
Genomic Map