Assay Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107002.1 | NP_001100472.1 | 5 - 6 | 812 | 57 | |
| XM_006246265.2 | XP_006246327.1 | 5 - 6 | 1024 | 57 | ||
| XM_006246266.2 | XP_006246328.1 | 5 - 6 | 825 | 57 | ||
| XM_006246267.2 | XP_006246329.1 | 5 - 6 | 819 | 57 | ||
| XM_006246268.1 | XP_006246330.1 | 5 - 6 | 787 | 57 | ||
| XM_006246270.2 | XP_006246332.1 | 5 - 6 | 825 | 57 | ||
| XM_006246271.2 | XP_006246333.1 | 5 - 6 | 825 | 57 | ||
| GenBank mRNA | BC158796.1 | - | 5 - 6 | 788 | 7314654 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Sept8
Genomic Map