Assay Details
| Gene Symbol: Myod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176079.1 | NP_788268.1 | 2 - 3 | 913 | 143 | |
| GenBank mRNA | BC127480.1 | - | 2 - 3 | 893 | 8366406 | 143 |
More Information
Updated Mapping Information
Set Membership: |
|

Myod1
Genomic Map