Assay Details
| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005900.1 | NP_001005900.1 | 2 - 3 | 136 | 59 | |
| XM_006228785.2 | XP_006228847.1 | 2 - 3 | 400 | 59 | ||
| GenBank mRNA | AY247020.1 | - | 2 - 3 | 136 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hcst
Genomic Map