Assay Details
| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039037.1 | NP_001034126.1 | 16 - 17 | 1779 | 78 | |
| XM_006229258.2 | XP_006229320.1 | 16 - 17 | 1798 | 78 | ||
| XM_008759402.1 | XP_008757624.1 | 16 - 17 | 1777 | 78 | ||
| GenBank mRNA | AY886529.1 | - | 16 - 17 | 1779 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
metabolic process
magnesium ion transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
cation transmembrane transport
signal transducer activity
zinc ion binding
magnesium ion transmembrane transporter activity
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
signal transduction
metabolic process
magnesium ion transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
cation transmembrane transport
signal transducer activity
zinc ion binding
magnesium ion transmembrane transporter activity
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P250476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9169 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P250476 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P250476 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P250476 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P250476 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64344 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46995 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17765 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10707171 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707172 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707165 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707174 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707173 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707170 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707168 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707167 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707169 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707164 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707161 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10707156 | - | - |
| Rat Genome 230 2.0 Array | 1398455_at | - | - |
| Rat Genome 230 2.0 Array | 1398455_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398455_at | - | - |
| Affymetrix HT Rat Focus Array | 1398455_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100012467.rn.1 | TC0100001541.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100012469.rn.1 | TC0100001541.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100012468.rn.1 | TC0100001541.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398455_at | - | - |
| Rat Expression Set 230 | 1398455_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707156 | - | - |
| Rat Genome U34 Set | rc_AI112197_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21799852 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352178 | - | - |

Zdhhc13
Genomic Map