Assay Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271081.1 | NP_001258010.1 | 3 | 2219 | 90 | |
| XM_006229055.2 | XP_006229117.1 | 3 | 2271 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lrrc4b
Genomic Map