Assay Details
| Gene Symbol: Zfp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228696.2 | XP_006228758.1 | 1 - 2 | 261 | 77 | |
| XM_006228697.2 | XP_006228759.1 | 1 - 2 | 261 | 77 | ||
| XM_006228698.2 | XP_006228760.1 | 2 - 3 | 584 | 77 | ||
| Gene Symbol: LOC102547413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759291.1 | XP_008757513.1 | 1 - 2 | 261 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp30
Genomic Map