Assay Details
| Gene Symbol: Ankrd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271264.1 | NP_001258193.1 | 22 - 23 | 2309 | 59 | |
| XM_008759172.1 | XP_008757394.1 | 22 - 23 | 2342 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd27
Genomic Map