Assay Details
| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238295.2 | XP_006238357.1 | 666 | 159 | ||
| GenBank mRNA | CO557703.1 | - | 253 | 159 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238295.2 | XP_006238357.1 | 666 | 159 | ||
| GenBank mRNA | CO557703.1 | - | 253 | 159 | ||
Set Membership: |
|