Assay Details
| Gene Symbol: Rbm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105898.1 | NP_001099368.1 | 1 - 2 | 161 | 87 | |
| XM_006247994.2 | XP_006248056.1 | 1 - 2 | 202 | 87 | ||
| XM_006247995.2 | XP_006248057.1 | 1 - 2 | 202 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm11
Genomic Map