Assay Details
| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107690.1 | NP_001101160.1 | 7 - 8 | 1381 | 112 | |
| XM_006232656.1 | XP_006232718.1 | 7 - 8 | 1844 | 112 | ||
| XM_006232664.1 | XP_006232726.1 | 7 - 8 | 1790 | 112 | ||
| XM_008761161.1 | XP_008759383.1 | 6 - 7 | 1979 | 112 | ||
| XM_008761162.1 | XP_008759384.1 | 6 - 7 | 1976 | 112 | ||
| XM_008761163.1 | XP_008759385.1 | 6 - 7 | 1967 | 112 | ||
| XM_008761164.1 | XP_008759386.1 | 6 - 7 | 1979 | 112 | ||
| XM_008761165.1 | XP_008759387.1 | 6 - 7 | 1937 | 112 | ||
| XM_008761166.1 | XP_008759388.1 | 6 - 7 | 1934 | 112 | ||
| XM_008761167.1 | XP_008759389.1 | 7 - 8 | 1832 | 112 | ||
| XM_008761168.1 | XP_008759390.1 | 7 - 8 | 1829 | 112 | ||
| XM_008761169.1 | XP_008759391.1 | 7 - 8 | 1802 | 112 | ||
| GenBank mRNA | FQ221099.1 | - | 7 - 8 | 1384 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam189b
Genomic Map