Assay Details
| Gene Symbol: Mansc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109603.2 | NP_001103073.1 | 1 - 2 | 222 | 67 | |
| XM_006237537.2 | XP_006237599.1 | 1 - 2 | 349 | 67 | ||
| XM_006237538.2 | XP_006237600.1 | 1 - 2 | 354 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mansc1
Genomic Map