Assay Details
| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001059437.5 | XP_001059437.3 | 4 - 5 | 1394 | 104 | |
| GenBank mRNA | BC158892.1 | - | 4 - 5 | 734 | 104 | |
More Information
Set Membership: |
|

Mbd4
Genomic Map